Product Name
Metastin, KISS-1 68-121 ≥95% (HPLC)
Product Number
SCP0186
Product Brand
SIGMA
CAS Number
Molecular Weight
5857.43
Test | Specification |
---|---|
Form | lyophilized powder |
Appearance | White to off-white |
Peptide Content | > or = to 75% |
Purity by HPLC | > or = to 95% |