Product Name
Monoclonal Anti-ARID1B antibody produced in mouse clone 2D2, purified immunoglobulin, buffered aqueous solution
Product Number
WH0057492M1
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Cell lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~236 kDa using Hela NE lysate. |
Immunohistochemistry-Paraffin Embedded Tissue (IHC-P) | Antibody concentration: 3 ug/ml using human kidney tissue. |
Immunofluorescence (IF) | Antibody concentration: 10 ug/ml using HeLa cell. |
Sandwich ELISA | Detection limit for recombinant GST tagged ARID1B is approximately 0.03ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~37 kDa using immunogen protein lysate. |