Skip to Content
MilliporeSigma
2-Chloro-4-iodoaniline 97%

2-Chloro-4-iodoaniline

2-Chloro-4-iodoaniline 97%
Empirical Formula (Hill Notation):
C6H5ClIN
CAS Number:
Molecular Weight:
253.47

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing