
4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-hydroxybenzamide

Synonym(s):
Metoclopramide EP Impurity F
Empirical Formula (Hill Notation):
C13H20ClN3O2
CAS Number:
Molecular Weight:
285.77
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1