Skip to Content
MilliporeSigma
GF 109203X synthetic, ≥90% (HPLC)

Bisindolylmaleimide I

GF 109203X synthetic, ≥90% (HPLC)
Synonym(s):
Bisindolylmaleimide I, 3-(N-[Dimethylamino]propyl-3-indolyl)-4-(3-indolyl)maleimide, 3-[1-[3-(Dimethylamino)propyl]1H-indol-3-yl]-4-(1Hindol-3-yl)1H-pyrrole-2,5dione, InSolution Bisindolylmaleimide I
Empirical Formula (Hill Notation):
C25H24N4O2
CAS Number:
Molecular Weight:
412.48

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment