Skip to Content
MilliporeSigma
Buffered Sodium Chloride Peptone Solution Tween® 0.1%, bottle capacity 500 mL, bottle filling volume 450 mL, closure type, Blue screw cap with 3 loci, pack of 6 bottles

Buffered Sodium Chloride Peptone Solution

Buffered Sodium Chloride Peptone Solution Tween® 0.1%, bottle capacity 500 mL, bottle filling volume 450 mL, closure type, Blue screw cap with 3 loci, pack of 6 bottles

Products

Product Category
Brand
Physical Form
Agency
Chemical Composition
Capacity (mL)
Quality Segment
Closure Type
Application
Package Size
Sterility
Sterilization Method
Technique
Showing 1-20 of 29
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2