
Meat Peptone for Biotechnology and Microbiology

Synonym(s):
Peptone from animal tissue
CAS Number:
EC Number:
Products
Product Category
Biological Source
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing