Skip to Content
MilliporeSigma
Methocarbamol-D3 solution 100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Methocarbamol-D3 solution

Methocarbamol-D3 solution 100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®
Empirical Formula (Hill Notation):
C11H12D3NO5
CAS Number:
Molecular Weight:
244.26

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique