
Octafluoro-2-butanone

Synonym(s):
Perfluoro-2-butanone
Linear Formula:
CF3CF2COCF3
CAS Number:
Molecular Weight:
216.03
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing