Skip to Content
MilliporeSigma
trans-α-Damascone ≥90%

trans-α-Damascone

trans-α-Damascone ≥90%
Synonym(s):
(±)-α-Damascone
Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30

Products

Product Category
Brand
Feature
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing