MilliporeSigma
GC Analysis of Trichloroanisole (TCA) and Precursors in Red Wine (Spiked) on SLB-5ms after SPME using 100 μm PDMS Fiber
GC Analysis of Trichloroanisole (TCA) and Precursors in Red Wine (Spiked) on SLB-5ms after SPME using 100 μm PDMS Fiber suitable for GC, application for SPME
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Polydimethylsiloxane (PDMS)
metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
accessory
Product No.
Description
Pricing
Vials, screw top with phenolic open-top cap, pre-assembled
volume 4 mL, clear glass vial, O.D. × H 15 mm × 45 mm, tan PTFE/silicone septum
related product
Product No.
Description
Pricing
Inlet Liner, Direct (SPME) Type, Straight Design (unpacked)
pkg of 1 ea
CONDITIONS
sample/matrix
1.5 mL red wine spiked with each analyte at 100 ng/L + 600 μL 5% potassium carbonate (K2CO3) + 240 mg sodium chloride (NaCl) + 60 μL acetic anhydride
SPME fiber
metal fiber coated with 100 μm PDMS (57928-U)
extraction
headspace, 50 °C for 30 min, with stirring
desorption process
250 °C for 3 min
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
50 °C (1 min), 25 °C/min to 280 °C
detector
ECD, 290 °C
carrier gas
helium, 1.5 mL/min, constant
liner
0.75 mm I.D. SPME