About This Item
Code UNSPSC :
41103406
Nomenclature NACRES :
NB.35
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aiderFabricant/nom de marque
Plas-Labs 800-HEATER
Description générale
The Plas-Labs catalyst heater has two primary functions for use in the Plas-Labs temperature and humidity glove box. The catalyst heater reduces trace amounts of oxygen and maintains incubation temperature levels of 35-37 °C. The internal atmosphere is drawn off the floor and up across heating elements and aluminized palladium pellets. Any oxygen present will react with your anaerobic gas mixture and be reduced to water vapor. When used in conjunction with the Plas-Labs temperature and humidity glove boxes with drying train attached, the vapor is then removed from the box as needed. CE compliant.
Informations légales
Still not finding the right product?
Explore all of our products under Catalyst heater for Plas-Labs temperature and humidity box