About This Item
Code UNSPSC :
31411800
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
Matériaux
EPDM rubber device (ethylene propylene diene monomer rubber)
PTFE
Niveau de qualité
Gamme de produits
NovAseptic®
Caractéristiques
hydrophobic (Surface)
Paramètres
—1 - 6 bar pressure range
—30 - 135 °C temp. range (Short term use)
Poids
0,01 kg (0.022 lb)
Application
The NovAseptic® valve is specifically designed for aseptic applications and complies with the most stringent cleanability and sterilization requirements. The focus on aseptic design is a significant feature for all valves. The NovAseptic® valve is designed for minimum dead leg, complete drainability and high chemical resistance.
Conditionnement
The Diaphragm is packaged in a closed box.
Autres remarques
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.
Informations légales
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany