Product Name
Neocuproine hydrate 99%
Product Number
121908
Marque du produit
ALDRICH
CAS Number
654054-57-6
Poids moléculaire
208.26 (anhydrous basis)
Test | Caractéristique |
---|---|
Appearance (Color) | Conforms |
White to off-white | |
Appearance (Form) | Conforms |
Powder | |
Infrared spectrum | Conforms with Structure |
Water (by Karl Fischer) | ≤10.00 % |
Purity (HPLC) | ≥98.50 % |
Solubility (Turbidity) | Conforms |
Clear | |
Solubility (Color) | Conforms |
Colorless to light yellow | |
Carbon (dry basis) | 79.50 - 82.00 % |
Nitrogen | 13.10 - 13.80 % |
Purity (TLC) | ≥99.00 % |
Consistent with 99% |