Product Name
4H-Pyran-4-one ≥98%
Product Number
177229
Marque du produit
ALDRICH
CAS Number
108-97-4
Poids moléculaire
96.08
Test | Caractéristique |
---|---|
APPEARANCE | WHITE TO YELLOW TO ORANGE CRYSTALS OR SOLID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 98.0% (MINIMUM) |
APPROVED DECEMBER 15, 2007 RJM |