Product Name
4-Ethylbenzoic acid 99%
Product Number
191280
Marque du produit
ALDRICH
CAS Number
619-64-7
Poids moléculaire
150.17
Test | Caractéristique |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER AND/OR CHUNKS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE AND STANDARD AS |
ILLUSTRATED ON PAGE 193C OF EDITION I, | |
VOLUME 2 OF "THE ALDRICH LIBRARY OF FT-IR | |
SPECTRA". | |
TITRATION | 98.5% - 101.5% (WITH NAOH) |
GAS LIQUID | 98.5% (MINIMUM) |
APPROVED AUGUST 25, 1999 JSB |