Product Name
1,1,3,3-Tetramethylbutyl isocyanide technical grade
Product Number
226491
Marque du produit
ALDRICH
CAS Number
14542-93-9
Poids moléculaire
139.24
Test | Caractéristique |
---|---|
APPEARANCE | COLORLESS TO YELLOW LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
OR | |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 94.0% (MINIMUM) |
APPROVED JUNE 21, 2007 RJM |