Product Name
2,2-Dimethyl-1-phenyl-1-propanol 99%
Product Number
337048
Marque du produit
ALDRICH
CAS Number
3835-64-1
Poids moléculaire
164.24
Test | Caractéristique |
---|---|
Appearance (Color) | White |
Appearance (Form) | Conforms to Requirements |
Solid to Powder and/or Chunks | |
Initial Melting Point | ≥41.0 °C |
Final Melting Point | ≤46.0 °C |
Infrared spectrum | Conforms to Structure |
Purity (GC) | ≥98.5 % |