Product Name
Fmoc-Met-OH ≥98.0% (HPLC)
Product Number
47634
Marque du produit
ALDRICH
CAS Number
71989-28-1
Poids moléculaire
371.45
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 98.0 % |
SPECIFIC ROTATION (20/D) | -31.0 - -28.0 Degrees |
CONCENTRATION | C=1 IN DIMETHYL FORMAMIDE |
CARBON CONTENT | 64.67 % (THEORY) |
HYDROGEN CONTENT | 5.70 % (THEORY) |
NITROGEN CONTENT | 3.77 % (THEORY) |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |