Product Name
9H-Carbazole-9-ethanol 95%
Product Number
479748
Marque du produit
ALDRICH
CAS Number
1484-14-6
Poids moléculaire
211.26
Test | Caractéristique |
---|---|
APPEARANCE | WHITE TO YELLOW TO TAN POWDER OR CRYSTALS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 74.8%-84.4% |
NITROGEN 6.2%- 7.0% | |
GAS LIQUID | 94.0% |
APPROVED AUGUST 4, 2008 RJM |