Product Name
5-Bromo-1-pentanol technical, ≥80% (GC)
Product Number
670855
Marque du produit
ALDRICH
CAS Number
34626-51-2
Poids moléculaire
167.04
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Colorless to Very Dark Brown-Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 80 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |