Product Name
1-Azido-4-fluorobenzene solution ~0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)
Product Number
779253
Marque du produit
ALDRICH
CAS Number
3296-02-4
Poids moléculaire
137.11
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Faint Yellow to Yellow and Faint Green-Yellow to Green-Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (HPLC AREA %) | ≥ 95.0 % |
DENSITY D20/4 | 0.770 - 0.790 |
WATER | ≤ 2.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
REMARKS | CONCENTRATION ~0.5M IN TBME |
RECOMMENDED RETEST PERIOD | 24 MONTHS |