Product Name
Phosphazene base P4-t-Bu solution ~0.8 M in hexane
Product Number
79421
Marque du produit
ALDRICH
CAS Number
111324-04-0
Poids moléculaire
633.72
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Colorless to Very Faint Yellow and Colorless to Very Faint Brown-Yellow |
APPEARANCE (FORM) | Liquid |
TITRATION (METHOD) | - |
TITRATION (MOLARITY) | 0.75 - 0.85 mol/l |
DENSITY D20/4 | 0.850 - 0.875 |
REFRACTIVE INDEX N20/D | 1.430 - 1.460 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
PHOSPHORUS 31 NMR SPECTRUM | CONFORMS TO STRUCTURE |