Product Name
Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate ≥98.0% (CHN)
Product Number
85971
Marque du produit
ALDRICH
CAS Number
207683-26-9
Poids moléculaire
411.28
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | White to Faint Beige |
APPEARANCE (FORM) | Powder or Crystals |
ASSAY (ELEMENTAL ANALYSIS) | ≥ 98.0 |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 1G IN 10ML ACETONITRILE |
CARBON CONTENT | 37.96 % (theory) |
NITROGEN CONTENT | 10.22 % (theory) |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |