Product Name
4-Acetylbutyric acid 97%
Product Number
A13204
Marque du produit
ALDRICH
CAS Number
3128-06-1
Poids moléculaire
130.14
Test | Caractéristique |
---|---|
APPEARANCE | COLORLESS TO DARK YELLOW LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5%-103.5% (WITH NAOH) |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED SEPTEMBER 4, 2008 RJM |