Product Name
2,6-Dibromopyridine 98%
Product Number
D43115
Marque du produit
ALDRICH
CAS Number
626-05-1
Poids moléculaire
236.89
Test | Caractéristique |
---|---|
APPEARANCE | WHITE TO GREY TO BEIGE POWDER, CRYSTALS OR |
CRYSTALLINE POWDER | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 97.5-102.5% (WITH AGNO3 AFTER O2 COMBUSTION) |
GAS LIQUID | 97.5% (MINIMUM) |
APPROVED JUNE 28, 2008 RJM |