Product Name
Phenoxazine 97%
Product Number
P14858
Marque du produit
ALDRICH
CAS Number
135-67-1
Poids moléculaire
183.21
Test | Caractéristique |
---|---|
APPEARANCE | TAN TO GREY POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | 75.9% - 81.4% CARBON |
7.4% - 7.9% NITROGEN | |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED FEBRUARY 21, 2006 JSB |