Product Name
Salicylhydroxamic acid 99%
Product Number
S607
Marque du produit
ALDRICH
CAS Number
89-73-6
Poids moléculaire
153.14
Test | Caractéristique |
---|---|
APPEARANCE | WHITE TO PINK POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 98.5%-101.5% (WITH NAOH) |
OR | |
MISCELLANEOUS ASSAYS | 98.5%-101.5% (TITRATION WITH TBAH) |
HIGH PRESSURE LIQUID | 98.5% (MINIMUM) |
REVISED FEBRUARY 29, 2008 RJM |