Product Name
5-Fluorouracil analytical standard
Product Number
03738
Marque du produit
SIAL
CAS Number
51-21-8
Poids moléculaire
130.08
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 99.0 % |
WATER | ≤ 0.5 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |