Product Name
Geranyl pyrophosphate lithium salt analytical standard
Product Number
19533
Marque du produit
SIAL
CAS Number
21141-43-5
Poids moléculaire
314.21 (free acid basis)
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder |
PURITY (HPLC AREA %) | ≥ 95.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
PHOSPHORUS 31 NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |