Product Name
Dichlormid PESTANAL®, analytical standard
Product Number
33613
Marque du produit
SIAL
CAS Number
37764-25-3
Poids moléculaire
208.09
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Colorless to Yellow to Brown-Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (HPLC AREA %) | ≥ 98.0 % |
REFRACTIVE INDEX N20/D | 1.5005 - 1.5045 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |