Product Name
2-NP-AMOZ VETRANAL®, analytical standard
Product Number
33869
Marque du produit
SIAL
CAS Number
183193-59-1
Poids moléculaire
334.33
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | White to Yellow |
APPEARANCE (FORM) | Powder or Crystals or Flakes |
PURITY (HPLC AREA %) | ≥ 99.0 % |
MELTING POINT | 144 - 150 C |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
RESIDUAL SOLVENTS | ≤ 1.0 % |
EXPIRY PERIOD | 36 MONTHS |