Product Name
T-2 Toxin-13C24 solution ~25 μg/mL in acetonitrile, analytical standard
Product Number
33892
Marque du produit
SIAL
CAS Number
Poids moléculaire
490.35
Test | Caractéristique |
---|---|
CONCENTRATION COMP1 | ~ 25 ug/ml |
MEASURING TOLERANCE P± | ≤ 2.5 ug/ml |
SOLVENT | ACETONITRILE |
PURITY (HPLC) COMP1 | ≥ 97.0 % |
MEASURING TOLERANCE P± COMP1 | ≤ 1.0 % |