Product Name
Dotetracontane analytical standard
Product Number
44250
Marque du produit
SIAL
CAS Number
7098-20-6
Poids moléculaire
591.13
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Crystals or Flakes |
PURITY (GC AREA %) | ≥ 98.0 % |
MELTING POINT | 83 - 86 C |
CARBON CONTENT | 85.34 % (Theory) |
HYDROGEN CONTENT | 14.66 % (Theory) |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |