Product Name
Sudan Red 7B analytical standard
Product Number
53373
Marque du produit
SIAL
CAS Number
6368-72-5
Poids moléculaire
379.46
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Red to Very Dark Red and Red-Brown and Very Dark Red-Brown |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 96.0 % |
IDENTIFICATION (LC-MS) | CORRESPONDS TO REQUIREMENTS |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 36 MONTHS |