Product Name
Electrolyte solution nonaqueous, 2 M LiCl in ethanol
Product Number
56754
Marque du produit
SIAL
CAS Number
7447-41-8
Poids moléculaire
42.39
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
TITRATION (METHOD) | -- |
TITRATION (MOLARITY) | 1.5 - 2.4 mol/l |
MISCELLANEOUS TESTS | ref.el. 6.0724.140/reference electrolyte/electrolyte; Ag/AgCl: mV 250 +/- 20 (25.0 C +/- 0.2 C) |
RECOMMENDED RETEST PERIOD | 57 MONTHS |