Product Name
Methyl nonanoate analytical standard
Product Number
76368
Marque du produit
SIAL
CAS Number
1731-84-6
Poids moléculaire
172.26
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 99.8 % |
REFRACTIVE INDEX N20/D | 1.421 - 1.423 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |