Product Name
Piperitone analytical standard
Product Number
79899
Marque du produit
SIAL
CAS Number
89-81-6
Poids moléculaire
152.23
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Colorless to Dark Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 98.0 % (mixture of enantiomers, predominantly (-)-enantiomer) |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |