Product Name
N-oxyde de N,N-diméthyldodécylamine BioXtra, ≥99.0% (NT)
Product Number
40234
Marque du produit
SIGMA
CAS Number
1643-20-5
Poids moléculaire
229.40
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder |
TITRATION (NT) HCLO4 0.1M | 99.0 - 101.0 % |
THIN LAYER CHROMATOGR. | CORRESPONDS TO STANDARD |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 0.5G IN 10ML METHANOL |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
TRACES PEROXIDE (AS H2O2) | ≤ 300 mg/kg |
METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
CALCIUM (ICP) | ≤ 10 mg/kg |
CADMIUM (ICP) | ≤ 5 mg/kg |
COBALT (ICP) | ≤ 5 mg/kg |
CHROMIUM (ICP) | ≤ 5 mg/kg |
COPPER (ICP) | ≤ 5 mg/kg |
IRON (ICP) | ≤ 5 mg/kg |
POTASSIUM (ICP) | ≤ 50 mg/kg |
MAGNESIUM (ICP) | ≤ 5 mg/kg |
MANGANESE (ICP) | ≤ 5 mg/kg |
SODIUM (ICP) | ≤ 50 mg/kg |
NICKEL (ICP) | ≤ 5 mg/kg |
LEAD (ICP) | ≤ 5 mg/kg |
ZINC (ICP) | ≤ 5 mg/kg |
CHLORIDE (CL) | ≤ 100 mg/kg |
SULFATE (SO4) | ≤ 50 mg/kg |
RECOMMENDED RETEST PERIOD | 54 MONTHS |