Product Name
Carbonate de sodium BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Product Number
71345
Marque du produit
SIGMA
CAS Number
497-19-8
Poids moléculaire
105.99
Test | Caractéristique |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Crystals |
TITRATION (T) HCL 1M | 99.5 - 101.0 % (calc. to the dried substance) |
LOSS ON DRYING | ≤ 1.0 % |
METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
ALUMINIUM (ICP) | ≤ 5 mg/kg |
BARIUM (ICP) | ≤ 5 mg/kg |
BISMUTH (ICP) | ≤ 5 mg/kg |
CALCIUM (ICP) | ≤ 50 mg/kg |
CADMIUM (ICP) | ≤ 5 mg/kg |
COBALT (ICP) | ≤ 5 mg/kg |
CHROMIUM (ICP) | ≤ 5 mg/kg |
COPPER (ICP) | ≤ 5 mg/kg |
IRON (ICP) | ≤ 5 mg/kg |
POTASSIUM (ICP) | ≤ 50 mg/kg |
LITHIUM (ICP) | ≤ 5 mg/kg |
MAGNESIUM (ICP) | ≤ 50 mg/kg |
MANGANESE (ICP) | ≤ 5 mg/kg |
MOLYBDENUM (ICP) | ≤ 5 mg/kg |
NICKEL (ICP) | ≤ 5 mg/kg |
LEAD (ICP) | ≤ 5 mg/kg |
STRONTIUM (ICP) | ≤ 5 mg/kg |
ZINC (ICP) | ≤ 5 mg/kg |
ARSENIC TRACES (MHS-AAS) | ≤ 0.1 mg/kg |
TOTAL SULFUR AS SO4 (ICP) | ≤ 50 mg/kg |
VARIOUS GUARANTEE LABEL TRACES (ICP-OES) | Phosphate, Silikate (as SiO(2)) ≤ 50 mg/kg |
CHLORIDE (CL) | ≤ 50 mg/kg |
TOTAL NITROGEN | ≤ 10 mg/kg |
SOLUBILITY (METHOD) | 1M water |
APPEARANCE (SOLUTION) | CLEAR COLORLESS |
PH (SOLUTION) | ~ 12.0 25C |
RESIDUE (FILTER TEST) | NO RESIDUE |
UV - ABS. AT 260 NM | ≤ 0.01 |
UV - ABS. AT 280 NM | ≤ 0.01 |
RECOMMENDED RETEST PERIOD | 72 MONTHS |