Product Name
Anti-ANO8 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Product Number
HPA049206
Marque du produit
SIGMA
CAS Number
Poids moléculaire
Test | Caractéristique |
---|---|
Protein Array | pass |
Immunohistochemistry Dilution | 1:50 - 1:200 |
Immunohistochemistry Antibody Retrieval Method | HIER pH6 |
Concentration | 0.1 mg/mL |