Product Name
Anti-GBA antibody produced in rabbit purified immunoglobulin, buffered aqueous solution
Product Number
SAB1410509
Marque du produit
SIGMA
CAS Number
Poids moléculaire
Test | Caractéristique |
---|---|
Appearance | Report result |
Concentration | Report result |
Western Blotting (Transfected lysate) | Antibody concentration: 0.1-5µg/ml. Specific band of ~59.7kDa using GBA transfected 293T lysate. |