Product Name
Anti-APOB antibody produced in goat affinity isolated antibody, buffered aqueous solution
Product Number
SAB2500080
Marque du produit
SIGMA
CAS Number
Poids moléculaire
Test | Caractéristique |
---|---|
Appearance | colorless liquid |
Concentration | 100µg/200µL |
ELISA (Peptide) | Antibody detection limit dilution 1:32000. |
Western Blotting (Cell lysate) | Bands corresponding to the expected sizes of ApoB100 and ApoB48 were detected in Human serum samples from five different individuals. Recommended concentration: 0.3-1 µg/ml. |
Immunohistochemistry - Paraffin Embedded Tissue | Not Verified |
Immunofluorescence | Not Verified |
Immunoprecipitation | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |