Product Name
Monoclonal Anti-Hsc70 (Hsp73) antibody produced in mouse clone N27F3-4, purified immunoglobulin
Product Number
SAB3701436
Marque du produit
SIGMA
CAS Number
Poids moléculaire
Test | Caractéristique |
---|---|
Appearance | Report result |
Concentration | Report result |
ELISA | 1:10,000 - 1:50,000 |
Immunohistochemistry | 1:200 - 1:1,000 |
Western Blot | 1:500 - 1:2,000 |