Product Name
Anti-JAK2 (Ab-570) antibody produced in rabbit affinity isolated antibody
Product Number
SAB4300587
Marque du produit
SIGMA
CAS Number
Poids moléculaire
Test | Caractéristique |
---|---|
Appearance | colorless liquid |
Concentration | 1.0 mg/ml |
ELISA | Not Determined |
Western Blotting | Not Determined |
Immunohistochemistry | 1:50-1:100 |
Immunofluorescence | Not Determined |
Immunoprecipitation | Not Determined |
Flow Cytometry | Not Determined |
Chromatin Immunoprecipitation | Not Determined |