Product Name
Anti-ADCY3 antibody produced in rabbit affinity isolated antibody
Product Number
SAB4301157
Marque du produit
SIGMA
CAS Number
Poids moléculaire
Test | Caractéristique |
---|---|
Appearance | Liquid |
Concentration | 2.5mg/mL |
ELISA | Not Determined |
Sandwhich ELISA | Not Determined |
Western Blotting (Cell Lysate) | 1:200-1:1000 |
Immunohistochemistry | 1:50-1:200 |
Immunofluorescence | Not Determined |
Immunoprecipitation | Not Determined |
Flow Cytopmetry | Not Determined |
Chromatin Immunoprecipitation | Not Determined |