Product Name
Monoclonal Anti-Thioredoxin (TRX) antibody produced in mouse clone 1H6H6, ascites fluid
Product Number
SAB5300166
Marque du produit
SIGMA
CAS Number
Poids moléculaire
Test | Caractéristique |
---|---|
Appearance | light yellow liquid |
Concentration | Not Available |
ELISA Antibody Concentration | Antibody concentration: 1/10000, using recombinant protein. |
Western Blotting (Recombinant Protein) | Antibody concentration: 1/500 - 1/2000. |
Western Blotting (Cell Lysate) | Antibody concentration: 1/500 - 1/2000. |
Immunohistochemistry | Not Determined |
Immunofluorescence | Not Determined |
Immunoprecipitation | Not Determined |
Flow Cytometry | Not Determined |
Chromatin Immunoprecipitation | Not Determined |