Product Name
D-Ile-Pro-Arg-AFC
Product Number
SCP0124
Marque du produit
SIGMA
CAS Number
Poids moléculaire
538.62
Test | Caractéristique |
---|---|
Form | lyophilized powder |
Appearance | White to off-white |
Peptide Content | > or = to 62% |
Purity by HPLC | > or = to 95% |