Product Name
FGF-23 human recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Product Number
SRP3039
Marque du produit
SIGMA
CAS Number
Poids moléculaire
Test | Caractéristique |
---|---|
Appearance | Report result |
Color | Report result |
Purity by SDS-Page | ≥95% |
Purity by HPLC | ≥95% |
Endotoxin Level | ≤ 1 EU/µg |
Biological Activity | Data not available |