Product Name
HGF mouse recombinant, expressed in Hi-5 Insect cells, ≥95% (SDS-PAGE), ≥95% (HPLC)
Product Number
SRP3300
Marque du produit
SIGMA
CAS Number
Poids moléculaire
Test | Caractéristique |
---|---|
Appearance | Report result |
color | Report result |
Purity by SDS-Page | ≥95% |
Purity by HPLC | ≥95% |
Endotoxin Level | ≤1 EU/µg |
Biological Activity | Determined by the dose-dependent stimulation of the proliferation of mouse IMCD3 cells using a concentration range of 10-20 ng/mL. |